missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7055797
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
188614-01-9
C10H9NO3S
223.25
MFCD00449104
UFSVRHNRNVVBJR-UHFFFAOYSA-N
699501
methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
COC(=O)C1=CC2=C(SCC(=O)N2)C=C1
This item is not returnable. View return policy
188614-01-9
C10H9NO3S
223.25
MFCD00449104
UFSVRHNRNVVBJR-UHFFFAOYSA-N
699501
methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
COC(=O)C1=CC2=C(SCC(=O)N2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 97°C to 99°C
Quantity 1 g
Beilstein 14311523
Synonym methyl 3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate, methyl-3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate, methyl 3-oxo-3,4-dihydro-2h-benzo b 1,4 thiazine-6-carboxylate, methyl 3-oxo-2h-1,4-benzothiazine-6-carboxylate, methyl 3,4-dihydro-3-oxo-2h-benzo b 1,4 thiazine-6-carboxylate, methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate, pubchem13044, ksc497g3b, methyl-3-oxo-3,4-dihydro-2h-1,, methyl3-oxo-2h-1,4-benzothiazine-6-carboxylate
Formula Weight 223.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'