Få mere at vide
Methyl 3-Mercaptobenzoate, ~85%, TRC
Artikelnummer. 30464716
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464716 1 g
30464496 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30464716 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3207301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72886-42-1
Molekylær formel C8 H8 O2 S
Molekylvægt (g/mol) 168.21
IUPAC navn methyl 3-sulfanylbenzoate
SMIL COC(=O)c1cccc(S)c1
Tekniske data
Kemisk navn eller materiale Methyl 3-Mercaptobenzoate
Mængde 1 g
InChI formel InChI=1S/C8H8O2S/c1-10-8(9)6-3-2-4-7(11)5-6/h2-5,11H,1H3
Formel vægt 168.0245