Få mere at vide
Methyl 3-(Hydroxymethyl)Benzoate, TRC
Artikelnummer. 30482251
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482251 10 g
30476391 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482251 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26555510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 3-(Hydroxymethyl)Benzoate
CAS 67853-03-6
Molekylær formel C9H10O3
Mængde 10 g
Synonym Methyl m-Hydroxymethylbenzoate, 3-(Methoxycarbonyl)benzyl Alcohol
InChI formel InChI=1S/C9H10O3/c1-12-9(11)8-4-2-3-7(5-8)6-10/h2-5,10H,6H2,1H3
SMIL COC(=O)c1cccc(CO)c1
IUPAC navn methyl 3-(hydroxymethyl)benzoate
Molekylvægt (g/mol) 166.17
Formel vægt 166.063