Få mere at vide
Methyl 3-hydroxy-4,5-dimethoxybenzoate, TRC
Artikelnummer. 30473240
Skift visning
Mængde:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473240 500 mg
30483347 2.5 g
30469846 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30473240 Leverandør Toronto Research Chemicals Leverandørnr. TRCM780505500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83011-43-2
Molekylær formel C10H12O5
Molekylvægt (g/mol) 212.2
Synonym Methyl 4,5-dimethoxy-3-hydroxybenzoate
IUPAC navn methyl 3-hydroxy-4,5-dimethoxybenzoate
SMIL COC(=O)c1cc(O)c(OC)c(OC)c1
Tekniske data
Kemisk navn eller materiale Methyl 3-hydroxy-4,5-dimethoxybenzoate
Mængde 500 mg
InChI formel InChI=1S/C10H12O5/c1-13-8-5-6(10(12)15-3)4-7(11)9(8)14-2/h4-5,11H,1-3H3
Formel vægt 212.0685