missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chlorosulfonyl-4-(isopropylsulfonyl)thiophene-2-carboxylate, 95%, Thermo Scientific™

Catalog Number 15488845
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
423768-47-2
C9H11ClO6S3
346.81
MFCD03407370
FHFIRKJBLQUUAD-UHFFFAOYSA-N
methyl 3-chlorosulfonyl-4-isopropylsulfonyl-2-thiophenecarboxylate, methyl 3-chlorosulfonyl-4-propane-2-sulfonyl thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-chlorosulfonyl-4-1-methylethyl sulfonyl-, methyl ester, methyl 3-chlorosulfonyl-4-isopropylsulfonyl thiophene-2-carboxylate, methyl 3-chlorosulfonyl-4-methylethyl sulfonyl thiophene-2-carboxylate
2779818
methyl 3-chlorosulfonyl-4-propan-2-ylsulfonylthiophene-2-carboxylate
COC(=O)C1=C(C(=CS1)S(=O)(=O)C(C)C)S(Cl)(=O)=O
This item is not returnable. View return policy
423768-47-2
C9H11ClO6S3
346.81
MFCD03407370
FHFIRKJBLQUUAD-UHFFFAOYSA-N
methyl 3-chlorosulfonyl-4-isopropylsulfonyl-2-thiophenecarboxylate, methyl 3-chlorosulfonyl-4-propane-2-sulfonyl thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-chlorosulfonyl-4-1-methylethyl sulfonyl-, methyl ester, methyl 3-chlorosulfonyl-4-isopropylsulfonyl thiophene-2-carboxylate, methyl 3-chlorosulfonyl-4-methylethyl sulfonyl thiophene-2-carboxylate
2779818
methyl 3-chlorosulfonyl-4-propan-2-ylsulfonylthiophene-2-carboxylate
COC(=O)C1=C(C(=CS1)S(=O)(=O)C(C)C)S(Cl)(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 115°C to 118°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 346.83
Percent Purity 95%
Chemical Name or Material Methyl 3-chlorosulfonyl-4-(isopropylsulfonyl)thiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'