missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-benzoylpropionate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.
Solubility
Soluble in Chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.
Solubility
Soluble in Chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
Specifications
Boiling Point | 172°C to 174°C (10 mmHg) |
Refractive Index | 1.524 |
Quantity | 5 g |
Beilstein | 2211627 |
Solubility Information | Soluble in Chloroform. |
Formula Weight | 192.22 |
Percent Purity | 98% |
Chemical Name or Material | Methyl 3-benzoylpropionate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'