missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-benzoylpropionate, 98%, Thermo Scientific Chemicals

Catalog Number 11484387
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
25333-24-8
C11H12O3
192.214
MFCD00014906
XVRCVKWYKYJEIG-UHFFFAOYSA-N
methyl 3-benzoylpropionate, 3-benzoylpropionic acid methyl ester, propionic acid, 3-benzoyl-, methyl ester, methyl 4-oxo-4-phenylbutyrate, benzenebutanoic acid, gamma-oxo-, methyl ester, benzenebutanoic acid, .gamma.-oxo-, methyl ester, benzenebutanoic acid, g-oxo-, methyl ester, acmc-209gja, methyl3-benzoylpropionate, methyl-3-benzoylpropionate
141190
methyl 4-oxo-4-phenylbutanoate
COC(=O)CCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
25333-24-8
C11H12O3
192.214
MFCD00014906
XVRCVKWYKYJEIG-UHFFFAOYSA-N
methyl 3-benzoylpropionate, 3-benzoylpropionic acid methyl ester, propionic acid, 3-benzoyl-, methyl ester, methyl 4-oxo-4-phenylbutyrate, benzenebutanoic acid, gamma-oxo-, methyl ester, benzenebutanoic acid, .gamma.-oxo-, methyl ester, benzenebutanoic acid, g-oxo-, methyl ester, acmc-209gja, methyl3-benzoylpropionate, methyl-3-benzoylpropionate
141190
methyl 4-oxo-4-phenylbutanoate
COC(=O)CCC(=O)C1=CC=CC=C1

Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.

Solubility
Soluble in Chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY
Boiling Point 172°C to 174°C (10 mmHg)
Refractive Index 1.524
Quantity 5 g
Beilstein 2211627
Solubility Information Soluble in Chloroform.
Formula Weight 192.22
Percent Purity 98%
Chemical Name or Material Methyl 3-benzoylpropionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'