Få mere at vide
Methyl 3-Aminobenzoate-2,4,5,6-d4, CDN
Artikelnummer. 30323845
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323845 0.25 g -
30316727 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323845 Leverandør CDN Leverandørnr. CDND71790.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Aminobenzoic acid ring-d4 methyl ester
CAS 911132-57-5
Molekylær formel C8 2H4 H5 N O2
Mængde 0.25 g
InChI formel InChI=1 S/C8H9NO2/c1-11-8(10)6-3-2-4-7(9)5-6/h2-5 H,9H2,1H3/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(N)c([2 H])c(C(=O)OC)c1[2 H]
IUPAC navn methyl 3-amino-2,4,5,6-tetradeuteriobenzoate
Molekylvægt (g/mol) 155.19
Formel vægt 155.0884 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity