missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, ≥95%, Thermo Scientific™

Catalog Number p-2704627
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
256353-38-5
C13H10F3NO2S
301.28
MFCD00832742
FYUVISRLFPIWPP-UHFFFAOYSA-N
methyl 3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl 3-amino-4-phenyl-5-trifluoromethyl-thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-phenyl-5-trifluoromethyl-, methyl ester, maybridge1_007879, methyl 3-amino-4-phenyl-5-trifuoromethyl thiophene-2carboxylate, methyl 3-azanyl-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl-3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, 3-amino-4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid methyl ester
2775320
methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
COC(=O)C1=C(N)C(=C(S1)C(F)(F)F)C1=CC=CC=C1
This item is not returnable. View return policy
256353-38-5
C13H10F3NO2S
301.28
MFCD00832742
FYUVISRLFPIWPP-UHFFFAOYSA-N
methyl 3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl 3-amino-4-phenyl-5-trifluoromethyl-thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-phenyl-5-trifluoromethyl-, methyl ester, maybridge1_007879, methyl 3-amino-4-phenyl-5-trifuoromethyl thiophene-2carboxylate, methyl 3-azanyl-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl-3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, 3-amino-4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid methyl ester
2775320
methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
COC(=O)C1=C(N)C(=C(S1)C(F)(F)F)C1=CC=CC=C1
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 301.29
Percent Purity 95%
Chemical Name or Material Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'