Få mere at vide
Methyl 3,6-dichloropyridine-2-carboxylate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Clopyralid-methyl |
| CAS | 1532-24-7 |
| Molekylær formel | C7 H5 Cl2 N O2 |
| Mængde | 250 mg |
| Synonym | Clopyralid-methyl, Picolinic acid, 3,6-dichloro-, methyl ester (7CI,8CI), 3,6-Dichloro-2-pyridinecarboxylic acid methyl ester, 3,6-Dichloropicolinic acid methyl ester, Clopyralid methyl ester, Clopyralid-methyl, Methyl 3,6-dichloro-2-pyridinecarboxylate, Methyl 3,6-dichloropicolinate |
| InChI formel | InChI=1S/C7H5Cl2NO2/c1-12-7(11)6-4(8)2-3-5(9)10-6/h2-3H,1H3 |
| SMIL | COC(=O)c1nc(Cl)ccc1Cl |
| IUPAC navn | methyl 3,6-dichloropyridine-2-carboxylate |
| Molekylvægt (g/mol) | 206.0261 |
| Formel vægt | 204.9697 |