missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 3,4-dichlorophenylacetate, 97+%, Thermo Scientific™

Artikelnummer. 11477074
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 g
10 g
50 g
Pakningsstørrelse:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11477074 50 g 50g
11457074 2 g 2g
11467074 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11477074 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B21518.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6725-44-6
Molekylær formel C9H8Cl2O2
Molekylvægt (g/mol) 219.061
MDL nummer MFCD00800680
InChI nøgle FWVOAIHAIWHHDM-UHFFFAOYSA-N
Synonym methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
PubChem CID 1268257
IUPAC navn methyl 2-(3,4-dichlorophenyl)acetate
SMIL COC(=O)CC1=CC(=C(C=C1)Cl)Cl

Tekniske data

Kemisk navn eller materiale Methyl 3,4-dichlorophenylacetate
Smeltepunkt 27°C to 30°C
Mængde 50 g
Formel vægt 219.07
Procent renhed ≥97%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.