Få mere at vide
Methyl 3,4-dichlorophenylacetate, 97+%, Thermo Scientific™
Artikelnummer. 11457074
Skift visning
Mængde:
2 g
10 g
50 g
Pakningsstørrelse:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11457074 2 g 2g
11467074 10 g 10g
11477074 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11457074 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B21518.04
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6725-44-6
Molekylær formel C9H8Cl2O2
Molekylvægt (g/mol) 219.061
MDL nummer MFCD00800680
InChI nøgle FWVOAIHAIWHHDM-UHFFFAOYSA-N
Synonym methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
PubChem CID 1268257
IUPAC navn methyl 2-(3,4-dichlorophenyl)acetate
SMIL COC(=O)CC1=CC(=C(C=C1)Cl)Cl
Tekniske data
Kemisk navn eller materiale Methyl 3,4-dichlorophenylacetate
Smeltepunkt 27°C to 30°C
Mængde 2 g
Formel vægt 219.07
Procent renhed ≥97%