Få mere at vide
Methyl 3-[[4-Chloro-2-(phenylamino)phenyl]methylamino]-3-oxopropanoate, TRC
Artikelnummer. 30133046
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133046 100 mg
30004885 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133046 Leverandør Toronto Research Chemicals Leverandørnr. TRCM295145100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2089061-92-5
Molekylvægt (g/mol) 332.78
IUPAC navn methyl 3-(2-anilino-4-chloro-N-methylanilino)-3-oxopropanoate
Tekniske data
Kemisk navn eller materiale Methyl 3-[[4-Chloro-2-(phenylamino)-phenyl]methylamino]-3-oxopropanoate
Molekylær formel C17 H17 Cl N2 O3
Mængde 100 mg
Synonym Methyl 3-[[4-Chloro-2-(phenylamino)-phenyl]methylamino]-3-oxopropanoate, Clobazam EP impurity F
InChI formel InChI=1S/C17H17ClN2O3/c1-20(16(21)11-17(22)23-2)15-9-8-12(18)10-14(15)19-13-6-4-3-5-7-13/h3-10,19H,11H2,1-2H3
SMIL COC(=O)CC(=O)N(C)c1ccc(Cl)cc1Nc2ccccc2
Formel vægt 332.0928