Få mere at vide
Methyl 3,4,5-Trimethoxycinnamate (>85%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 3,4,5-Trimethoxycinnamate |
| CAS | 20329-96-8 |
| Molekylær formel | C13 H16 O5 |
| Mængde | 50 mg |
| Synonym | 2-Propenoic acid, 3-(3,4,5-trimethoxyphenyl)-, methyl ester, (2E)- (9CI, ACI), 2-Propenoic acid, 3-(3,4,5-trimethoxyphenyl)-, methyl ester, (E)- (ZCI), Cinnamic acid, 3,4,5-trimethoxy-, methyl ester, (E)- (8CI), Methyl (E)-3,4,5-trimethoxycinnamate, Methyl 3,4,5-trimethoxy-trans-cinnamate, Methyl trans-3,4,5-trimethoxycinnamate, Methyl trans-3-(3,4,5-trimethoxyphenyl)acrylate, trans-3-(3,4,5-Tri-methoxyphenyl-3-Propenoic acid methyl ester |
| InChI formel | InChI=1S/C13H16O5/c1-15-10-7-9(5-6-12(14)17-3)8-11(16-2)13(10)18-4/h5-8H,1-4H3/b6-5+ |
| SMIL | COC(=O)\C=C\c1cc(OC)c(OC)c(OC)c1 |
| IUPAC navn | methyl (E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoate |
| Molekylvægt (g/mol) | 252.26 |
| Formel vægt | 252.0998 |