missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 3-(3-(Benzyloxy)phenyl)-3-oxopropanoate, TRC

Artikelnummer. 30479878
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479878 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30479878 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3292835MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 3-(3-(Benzyloxy)phenyl)-3-oxopropanoate
CAS 150356-53-9
Molekylær formel C17H16O4
Mængde 5 mg
Synonym beta-Oxo-3-(phenylmethoxy)-benzenepropanoic Acid Methyl Ester
InChI formel InChI=1S/C17H16O4/c1-20-17(19)11-16(18)14-8-5-9-15(10-14)21-12-13-6-3-2-4-7-13/h2-10H,11-12H2,1H3
SMIL COC(=O)CC(=O)c1cccc(OCc2ccccc2)c1
IUPAC navn methyl 3-oxo-3-(3-phenylmethoxyphenyl)propanoate
Molekylvægt (g/mol) 284.31
Formel vægt 284.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.