missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 3-(3’,4’-Dimethoxyphenyl)propenoate, TRC

Product Code. 30473982
Change view
Click to view available options
Mængde:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30473982 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473982 Supplier Toronto Research Chemicals Supplier No. TRCM3028702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 3-(3',4'-Dimethoxyphenyl)propenoate
CAS 5396-64-5
Molekylær formel C12 H14 O4
Mængde 2.5 g
Synonym Methyl 3,4-Dimethoxycinnamate, 3-(3,4-Dimethoxyphenyl)-2-propenoic Acid Methyl Ester, 3,4-O-Dimethylcaffeic Acid Methyl Ester, NSC 4331
InChI formel InChI=1S/C12H14O4/c1-14-10-6-4-9(8-11(10)15-2)5-7-12(13)16-3/h4-8H,1-3H3
SMIL COC(=O)C=Cc1ccc(OC)c(OC)c1
IUPAC navn methyl 3-(3,4-dimethoxyphenyl)prop-2-enoate
Molekylvægt (g/mol) 222.24
Formel vægt 222.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.