Få mere at vide
Methyl 3-[[(2RS)-2-Bromopropanoyl]amino]-4-methylthiophene-2-carboxylate (Bromo Compound), Mikromol
Beskrivelse
Methyl 3-[[(2RS)-2-Bromopropanoyl]amino]-4-methylthiophene-2-carboxylate (Bromo Compound), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 3-([(2RS)-2-Bromopropanoyl]amino)-4-methylthiophene-2-carboxylate |
| CAS | 1160919-46-9 |
| Analyt- eller komponentnavne | Methyl 3-([(2RS)-2-Bromopropanoyl]amino)-4-methylthiophene-2-carboxylate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H12 Br N O3 S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Articaine Hydrochloride Imp. J (EP), Articaine Imp. J (EP), Methyl 3-[[(2RS)-2-Bromopropanoyl]amino]-4-methylthiophene-2-carboxylate, Bromo Compound of Articaine, Articaine Hydrochloride Impurity J, Articaine Impurity J |
| InChI formel | InChI=1S/C25H30INO3.ClH/c1-4-7-11-23-24(19-10-8-9-12-21(19)30-23)25(28)18-13-14-22(20(26)17-18)29-16-15-27(5-2)6-3;/h8-10,12-14,17H,4-7,11,15-16H2,1-3H3;1H |
| Løsningstype | Neat |
| SMIL | COC(=O)c1scc(C)c1NC(=O)C(C)Br |
| Vis mere |