Få mere at vide
Methyl 3-(2-Ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate, TRC
Artikelnummer. 30471522
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30471522 2.5 mg 2.5mg
30477812 10 mg -
30475884 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30471522 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3167552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2518276-69-0
Molekylær formel C19H25N3O3
Molekylvægt (g/mol) 343.43
Synonym Remifentanil EP Impurity L
IUPAC navn methyl 3-(2-ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate
SMIL CCC1=NC(=O)C2(CCN(CCC(=O)OC)CC2)N1c3ccccc3
Tekniske data
Kemisk navn eller materiale Methyl 3-(2-Ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate
Mængde 2.5 mg
InChI formel InChI=1S/C19H25N3O3/c1-3-16-20-18(24)19(22(16)15-7-5-4-6-8-15)10-13-21(14-11-19)12-9-17(23)25-2/h4-8H,3,9-14H2,1-2H3
Formel vægt 343.1896