missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl (2R)-2-Amino-2-phenylacetate Hydrochloride, Mikromol™

Artikelnummer. 30049366
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049366 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30049366 Leverandør Mikromol Leverandørnr. MM0605.110025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl (2R)-2-Amino-2-phenylacetate Hydrochloride
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl (2R)-2-Amino-2-phenylacetate Hydrochloride
CAS 19883-41-1
Analyt- eller komponentnavne Methyl (2R)-2-Amino-2-phenylacetate Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C9 H11 N O2 . H Cl
Certificeringer/compliance ISO 9001
Synonym Benzeneacetic acid, α-amino-, methyl ester, hydrochloride (1:1), (αR)-, Benzeneacetic acid, α-amino-, methyl ester, hydrochloride, (R)-, Benzeneacetic acid, α-amino-, methyl ester, hydrochloride, (αR)- (9CI), Glycine, 2-phenyl-, methyl ester, hydrochloride, D- (8CI), ((R)-2-Methoxy-2-oxo-1-phenylethyl)ammonium chloride, (R)-(-)-2-Phenylglycine methyl ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, (R)-Methyl 2-amino-2-phenylacetate hydrochloride, (R)-Methyl 2-amino-2-phenylacetate hydrochloride, (R)-Phenylglycine methyl ester hydrochloride, D-Phenylglycine methyl ester hydrochloride, D-α-Aminophenylacetic acid methyl ester hydrochloride, Methyl (R)-amino(phenyl)acetate hydrochloride, Methyl (R)-phenylglycinate hydrochloride, Methyl D-phenylglycinate hydrochloride, Methyl D-α-phenylglycinate hydrochloride, Methyl D-α-phenylglycine hydrochloride
InChI formel InChI=1S/C9H11NO2.ClH/c1-12-9(11)8(10)7-5-3-2-4-6-7;/h2-6,8H,10H2,1H3;1H/t8-;/m1./s1
SMIL Cl.COC(=O)[C@H](N)c1ccccc1
IUPAC navn methyl (2R)-2-amino-2-phenyl-acetate;hydrochloride
Molekylvægt (g/mol) 201.65
Mængde 25 mg
Formel vægt 201.0557 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.