Få mere at vide
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate, TRC
Artikelnummer. 30473720
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473720 500 mg
30474684 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473720 Leverandør Toronto Research Chemicals Leverandørnr. TRCM330010500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57113-90-3
Molekylær formel C13H16N2O6
Molekylvægt (g/mol) 296.28
Synonym 2-[[(1,1-Dimethylethoxy)carbonyl]amino]-3-nitrobenzoic Acid Methyl Ester
IUPAC navn methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-nitrobenzoate
SMIL COC(=O)c1cccc(c1NC(=O)OC(C)(C)C)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate
Mængde 500 mg
InChI formel InChI=1S/C13H16N2O6/c1-13(2,3)21-12(17)14-10-8(11(16)20-4)6-5-7-9(10)15(18)19/h5-7H,1-4H3,(H,14,17)
Formel vægt 296.1008