Få mere at vide
Methyl (2-Oxopyrrolidin-1-yl)acetate, Mikromol™
Beskrivelse
Methyl (2-Oxopyrrolidin-1-yl)acetate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl (2-Oxopyrrolidin-1-yl)acetate |
| CAS | 59776-88-4 |
| Analyt- eller komponentnavne | Methyl (2-Oxopyrrolidin-1-yl)acetate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C7 H11 N O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Methyl (2-Oxopyrrolidin-1-yl)acetate, Piracetam Imp. B (EP), 2-Oxo-1-pyrrolidineacetic acid methyl ester, (2-Oxopyrrolidin-1-yl)acetic acid methyl ester, 1-(Methoxycarbonylmethyl)-2-pyrrolidone, 2-Ketopyrrolidine-1-acetic acid methyl ester, Methyl 2-(2-oxo-1-pyrrolidinyl)acetate, Methyl 2-oxo-1-pyrrolidineacetate, Methyl 2-pyrrolidinon-1-ylacetate, Methyl 2-pyrrolidone-1-acetate, Piracetam Impurity B |
| InChI formel | InChI=1S/C7H11NO3/c1-11-7(10)5-8-4-2-3-6(8)9/h2-5H2,1H3 |
| SMIL | COC(=O)CN1CCCC1=O |
| IUPAC navn | methyl 2-(2-oxopyrrolidin-1-yl)acetate |
| Vis mere |