Få mere at vide
Methyl 2-Oxo-1-pyrrolidineacetate, TRC
Artikelnummer. 30008858
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008858 5 g
30004081 10 g
17983875 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008858 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3210655G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59776-88-4
Molekylær formel C7 H11 N O3
Molekylvægt (g/mol) 157.17
Synonym Methyl (2-Oxopyrrolidin-1-yl)acetate, Piracetam Imp. B (EP), 2-Oxo-1-pyrrolidineacetic acid methyl ester, (2-Oxopyrrolidin-1-yl)acetic acid methyl ester, 1-(Methoxycarbonylmethyl)-2-pyrrolidone, 2-Ketopyrrolidine-1-acetic acid methyl ester, Methyl 2-(2-oxo-1-pyrrolidinyl)acetate, Methyl 2-oxo-1-pyrrolidineacetate, Methyl 2-pyrrolidinon-1-ylacetate, Methyl 2-pyrrolidone-1-acetate
IUPAC navn methyl 2-(2-oxopyrrolidin-1-yl)acetate
SMIL COC(=O)CN1CCCC1=O
Tekniske data
Kemisk navn eller materiale Methyl (2-Oxopyrrolidin-1-yl)acetate
Renhedsgrad noter HPLC
InChI formel InChI=1S/C7H11NO3/c1-11-7(10)5-8-4-2-3-6(8)9/h2-5H2,1H3
Mængde 5 g
Formel vægt 157.0739
Procent renhed >95%