missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 2-O-(Alpha-D-Mannopyranosyl)-Alpha-D-mannopyranoside, TRC

Artikelnummer. 30467111
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467111 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30467111 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3199801MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl 2-O-(a-D-Mannopyranosyl)-a-D-mannopyranoside
CAS 59571-75-4
Molekylær formel C13 H24 O11
Mængde 1 mg
InChI formel InChI=1S/C13H24O11/c1-21-13-11(9(19)7(17)5(3-15)23-13)24-12-10(20)8(18)6(16)4(2-14)22-12/h4-20H,2-3H2,1H3/t4-,5-,6-,7-,8+,9+,10+,11+,12-,13+/m1/s1
SMIL CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]2O
IUPAC navn (2R,3S,4S,5S,6R)-2-[(2S,3S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-methoxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvægt (g/mol) 356.32
Formel vægt 356.1319
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.