Learn More
Methyl 2-naphthoate, 97%, BLD Pharm™
Product Code. 30947976
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
250mg
25g
500g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947976 1 g 1g
30939640 500 g 500g
30935549 25 g 25g
30945545 100 g 100g
30939487 5 g 5g
30932249 250 mg 250mg
30930449 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30947976 Supplier BLD Pharm Supplier No. BD137761g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 2459-25-8
Molecular Formula C12H10O2
MDL Number MFCD00017345
Quantity 1 g
SMILES O=C(OC)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 186.21
Percent Purity 97%
Chemical Name or Material Methyl 2-naphthoate