missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(methylthio)pyrimidine-5-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | methyl 2-methylthio pyrimidine-5-carboxylate, methyl 2-methylsulfanyl pyrimidine-5-carboxylate, methyl 2-methylthiopyrimidine-5-carboxylate, methyl 2-methylthio-5-pyrimidinecarboxylate, 2-methylthio-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylic acid, 2-methylthio-, methyl ester, methyl2-methylthio pyrimidine-5-carboxylate, methyl 2-methylthio-5-pyrimidinearboxylate, methyl-2-methylthio pyrimidine-5-carboxylate, methyl 2-methylsulphanyl pyrimidine-5-carboxylate |
IUPAC Name | methyl 2-(methylsulfanyl)pyrimidine-5-carboxylate |
Formula Weight | 184.22 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-(methylthio)pyrimidine-5-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'