missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 2-Hydroxybenzoate-3,4,5,6-d4, TRC

Artikelnummer. 30469652
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469652 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469652 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3310261MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl Salicylate-3,4,5,6-D4
CAS 1219802-12-6
Molekylær formel C8H4D4O3
Mængde 1 mg
Synonym Methyl salicylate-3,4,5,6-D4, Methyl salicylate D4 (phenyl-D4), Methyl salicylate-d4, Methyl 2-hydroxybenzoate-3,4,5,6-d4, 2-Hydroxybenzoic-3,4,5,6-d4 acid methyl ester, Salicylic-3,4,5,6-d4 acid methyl ester
InChI formel InChI=1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3/i2D,3D,4D,5D
SMIL O=C(OC)C1=C([2H])C([2H])=C([2H])C([2H])=C1O
IUPAC navn methyl 2-hydroxybenzoate-3,4,5,6-d4
Molekylvægt (g/mol) 156.17
Formel vægt 156.07
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.