Få mere at vide
Methyl 2-Hydroxy-5-methylbenzoate, TRC
Artikelnummer. 30338045
Skift visning
Mængde:
1 g
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30338045 1 g -
30335309 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30338045 Leverandør Toronto Research Chemicals Leverandørnr. TRCB5853181G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 2-Hydroxy-5-methylbenzoate
CAS 22717-57-3
Molekylær formel C9H10O3
Mængde 1 g
Synonym 2-(Methoxycarbonyl)-4-methylphenol, 2-Hydroxy-5-methylbenzoic Acid Methyl Ester, Methyl 5-Methylsalicylate, NSC 165635
InChI formel InChI=1S/C9H10O3/c1-6-3-4-8(10)7(5-6)9(11)12-2/h3-5,10H,1-2H3
SMIL COC(=O)c1cc(C)ccc1O
IUPAC navn methyl 2-hydroxy-5-methylbenzoate
Molekylvægt (g/mol) 166.18
Formel vægt 166.063