missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-chloro-5-nitrobenzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
2.5 g
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl 2-chloro-5-nitrobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Methyl 2-chloro-5-nitrobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Melting Point | 70°C to 72°C |
Quantity | 2.5 g |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 215.59 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-chloro-5-nitrobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'