missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-chloro-4,4-dimethyl-3-oxopentanoate, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)3CC(O)CHClCO2CH3 |
Quantity | 1 g |
Synonym | 2-chloro-4,4-dimethyl-3-oxo-pentanoic acid methyl ester, methyl 2-chloro-4,4-dimethyl-3-oxo-pentanoate, pentanoic acid,2-chloro-4,4-dimethyl-3-oxo-, methyl ester, pentanoic acid, 2-chloro-4,4-dimethyl-3-oxo-, methyl ester |
Formula Weight | 192.64 |
Percent Purity | 90% |
Chemical Name or Material | Methyl 2-chloro-4, 4-dimethyl-3-oxopentanoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'