missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-6-fluorophenylacetate, 96%, Thermo Scientific™

Catalog Number p-7054339
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1427397-15-6
C9H8BrFO2
247.063
QXIUUACWTNNJID-UHFFFAOYSA-N
methyl 2-2-bromo-6-fluorophenyl acetate
100912176
methyl 2-(2-bromo-6-fluorophenyl)acetate
COC(=O)CC1=C(C=CC=C1Br)F
This item is not returnable. View return policy
1427397-15-6
C9H8BrFO2
247.063
QXIUUACWTNNJID-UHFFFAOYSA-N
methyl 2-2-bromo-6-fluorophenyl acetate
100912176
methyl 2-(2-bromo-6-fluorophenyl)acetate
COC(=O)CC1=C(C=CC=C1Br)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 247.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 2-bromo-6-fluorophenylacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'