missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-bromo-3-fluoropropionate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Boiling Point | 50°C to 54°C (8 mmHg) |
Quantity | 250 mg |
UN Number | UN3265 |
Solubility Information | Insoluble in water. |
Formula Weight | 184.99 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-bromo-3-fluoropropionate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'