missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 2-amino-5-methylthiophen-3-carboxylat, TRC

Artikelnummer. 30475212
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475212 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30475212 Leverandør Toronto Research Chemicals Leverandørnr. TRCM286865500MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Læg i indkøbskurv
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 2-Amino-5-methylthiophene-3-carboxylate
CAS 19369-53-0
Molekylær formel C7H9NO2S
Mængde 500 mg
Synonym 2-Amino-5-methyl-3-thiophenecarboxylic Acid Methyl Ester, 2-Amino-3-(methoxycarbonyl)-5-methylthiophene, 2-Amino-5-methylthiophene-3-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C7H9NO2S/c1-4-3-5(6(8)11-4)7(9)10-2/h3H,8H2,1-2H3
SMIL COC(=O)c1cc(C)sc1N
IUPAC navn methyl 2-amino-5-methylthiophen-3-carboxylat
Molekylvægt (g/mol) 171.22
Formel vægt 171.0354
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.