Få mere at vide
Methyl 2,5-Dihydroxycinnamate, TRC
Artikelnummer. 30477352
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30477352 10 mg
1 options
Artikelnummer. 30477352 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30270010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl (2E)-3-(2,5-Dihydroxyphenyl)-2-propenoate
CAS 123064-80-2
Molekylær formel C10 H10 O4
Mængde 10 mg
Synonym 2-Propenoic acid','3-(2,5-dihydroxyphenyl)-','methyl ester','(2E)-, 2-Propenoic acid','3-(2,5-dihydroxyphenyl)-','methyl ester','(E)-, Methyl grevillate, (E)-2,5-Dihydroxycinnamic acid methyl ester, Methyl (E)-2,5-dihydroxycinnamate, Methyl trans-2,5-dihydroxycinnamate, (E)-3-(2,5-Dihydroxyphenyl)-2-propenoic acid methyl ester, Methyl trans-3-(2,5-dihydroxyphenyl)acrylate, Methyl (2E)-3-(2,5-dihydroxyphenyl)-2-propenoate, 3-(2,5-Dihydroxyphenyl)-2-propenoic Acid Methyl Ester, Methyl 2,5-Dihydroxycinnamate
InChI formel InChI=1S/C10H10O4/c1-14-10(13)5-2-7-6-8(11)3-4-9(7)12/h2-6,11-12H,1H3/b5-2+
SMIL COC(=O)\C=C\c1cc(O)ccc1O
IUPAC navn methyl (E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
Molekylvægt (g/mol) 194.184
Formel vægt 194.0579