Learn More
Methyl 2,5-dichloronicotinate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 67754-03-4 |
| Molecular Formula | C7H5Cl2NO2 |
| MDL Number | MFCD03844857 |
| Quantity | 250 mg |
| SMILES | O=C(OC)C1=C(Cl)N=CC(Cl)=C1 |
| Molecular Weight (g/mol) | 206.03 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 2,5-dichloronicotinate |