missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(4-Bromophenyl)-2,2-dimethylacetate, TRC

Product Code. 30471120
Change view
Click to view available options
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30471120 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471120 Supplier Toronto Research Chemicals Supplier No. TRCM2943905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 2-(4-Bromophenyl)-2,2-dimethylacetate
CAS 154825-97-5
Molekylær formel C11 H13 Br O2
Mængde 5 g
InChI formel InChI=1S/C11H13BrO2/c1-11(2,10(13)14-3)8-4-6-9(12)7-5-8/h4-7H,1-3H3
SMIL COC(=O)C(C)(C)c1ccc(Br)cc1
IUPAC navn methyl 2-(4-bromophenyl)-2-methylpropanoate
Molekylvægt (g/mol) 257.12
Formel vægt 256.0099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.