Få mere at vide
Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Mikromol™
Artikelnummer. 17903186
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17903186 100 mg
17950706 25 mg
30462746 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17903186 Leverandør Mikromol Leverandørnr. MM0063.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
CAS 2157488-46-3
Analyt- eller komponentnavne Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H22 Cl N O4
Certificeringer/compliance ISO 17025
Synonym Bezafibrate Imp. C (EP), Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Bezafibrate Impurity C
InChI formel InChI=1S/C17H14N4O4/c1-25-17(24)21-16-19-13-7-4-11(8-14(13)20-16)15(23)10-2-5-12(6-3-10)18-9-22/h2-9H,1H3,(H,18,22)(H2,19,20,21,24)
SMIL COC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1
IUPAC navn methyl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Molekylvægt (g/mol) 375.85
Mængde 100 mg
Formel vægt 375.1237 g/mol