Få mere at vide
Methyl 2,3-Dichlorotrifluoropropionate, TRC
Artikelnummer. 30469253
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469253 50 mg
30469549 100 mg
30477279 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30469253 Leverandør Toronto Research Chemicals Leverandørnr. TRCM69618050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16329-89-8
Molekylær formel C4H3O2F3Cl2
Molekylvægt (g/mol) 210.96
Synonym Methyl 2,3-Dichloro-2,3,3-trifluoropropanoate, Methyl 2,3-Dichloro-2,3,3-trifluoropropionate, Methyl 2,3-Dichlorotrifluoropropionate
IUPAC navn methyl 2,3-dichloro-2,3,3-trifluoropropanoate
SMIL COC(=O)C(F)(Cl)C(F)(F)Cl
Tekniske data
Kemisk navn eller materiale Methyl 2,3-Dichlorotrifluoropropionate
Mængde 50 mg
InChI formel InChI=1S/C4H3Cl2F3O2/c1-11-2(10)3(5,7)4(6,8)9/h1H3
Formel vægt 209.9462