missing translation for 'onlineSavingsMsg'
Få mere at vide

Methyl 2,3,4-Tri-O-acetyl-Beta-D-glucuronic Acid Methyl Ester, TRC

Artikelnummer. 30480732
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30480732 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480732 Supplier Toronto Research Chemicals Supplier No. TRCM33081925MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methyl 2,3,4-Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
CAS 34213-34-8
Molekylær formel C14 H20 O10
Mængde 25 mg
Synonym β-D-Glucopyranosiduronic acid, methyl, methyl ester, 2,3,4-triacetate, Glucopyranosiduronic acid, methyl, methyl ester, triacetate (7CI), Glucopyranosiduronic acid, methyl, methyl ester, triacetate, β-D- (8CI), Glucosiduronic acid, methyl, methyl ester, triacetate (6CI), β-D-Glucopyranosiduronic acid, methyl, methyl ester, triacetate (9CI), Methyl 2,3,4-tri-O-acetyl-β-D-glucuronic acid methyl ester, Methyl 2,3,4-tri-O-acetyl-β-D-glucopyranosiduronic acid methyl ester
InChI formel InChI=1S/C14H20O10/c1-6(15)21-9-10(22-7(2)16)12(23-8(3)17)14(20-5)24-11(9)13(18)19-4/h9-12,14H,1-5H3/t9-,10-,11-,12+,14+/m0/s1
SMIL CO[C@@H]1O[C@@H]([C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C)C(=O)OC
IUPAC navn methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-methoxyoxane-2-carboxylate
Molekylvægt (g/mol) 348.3
Formel vægt 348.1056
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.