Learn More
Alfa Aesar™ Methyl 2-(2-chloroacetamido)-4-phenylthiophene-3-carboxylate, 96%
Brand: Alfa Aesar™ H33721.06
Additional Details : CAS Number : 356568-66-6 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
356568-66-6 | |
309.764 | |
QVZIDNLJEOTBPT-UHFFFAOYSA-N | |
1536902 | |
COC(=O)C1=C(SC=C1C2=CC=CC=C2)NC(=O)CCl |
C14H12ClNO3S | |
MFCD01917476 | |
methyl 2-chloroacetyl amino-4-phenylthiophene-3-carboxylate, methyl 2-2-chloroacetamido-4-phenylthiophene-3-carboxylate, methyl 2-2-chloroacetylamino-4-phenylthiophene-3-carboxylate, methyl 2-2-chloroacetyl amino-4-phenylthiophene-3-carboxylate | |
methyl 2-[(2-chloroacetyl)amino]-4-phenylthiophene-3-carboxylate |
Specifications
Methyl 2-(2-chloroacetamido)-4-phenylthiophene-3-carboxylate | |
5g | |
C14H12ClNO3S | |
methyl 2-chloroacetyl amino-4-phenylthiophene-3-carboxylate, methyl 2-2-chloroacetamido-4-phenylthiophene-3-carboxylate, methyl 2-2-chloroacetylamino-4-phenylthiophene-3-carboxylate, methyl 2-2-chloroacetyl amino-4-phenylthiophene-3-carboxylate | |
COC(=O)C1=C(SC=C1C2=CC=CC=C2)NC(=O)CCl | |
309.764 | |
309.77 |
107°C to 110°C | |
356568-66-6 | |
MFCD01917476 | |
QVZIDNLJEOTBPT-UHFFFAOYSA-N | |
methyl 2-[(2-chloroacetyl)amino]-4-phenylthiophene-3-carboxylate | |
1536902 | |
96% |