Learn More
Methyl 2,2,3,3,4,4,4-heptafluorobutanoate, 97%, BLD Pharm™
Product Code. 30958821
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30958821 5 g 5g
30954476 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30958821 Supplier BLD Pharm Supplier No. BD1394485g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 356-24-1
Molecular Formula C5H3F7O2
MDL Number MFCD00000433
Quantity 5 g
SMILES O=C(OC)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 228.07
Percent Purity 97%
Chemical Name or Material Methyl 2,2,3,3,4,4,4-heptafluorobutanoate