Få mere at vide
Methyl 2-(1-Hexyn-1-yl)benzoate, TRC
Artikelnummer. 30480039
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480039 250 mg
30476089 50 mg
30469259 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480039 Leverandør Toronto Research Chemicals Leverandørnr. TRCM335105250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 462637-40-7
Molekylær formel C14H16O2
Molekylvægt (g/mol) 216.28
Synonym 1-(2-Methoxycarbonylphenyl)-1-hexyne, 2-(1-Hexynyl)-benzoic Acid Methyl Ester
IUPAC navn methyl 2-(hex-1-yn-1-yl)benzoate
SMIL ClCC1=CC=C(SC2=CC=CC=C2)C=C1
Tekniske data
Kemisk navn eller materiale Methyl 2-(1-Hexyn-1-yl)benzoate
Mængde 250 mg
InChI formel InChI=1S/C14H16O2/c1-3-4-5-6-9-12-10-7-8-11-13(12)14(15)16-2/h7-8,10-11H,3-5H2,1-2H3
Formel vægt 216.12