Få mere at vide
Methyl (1S)-(+)-10-Camphorsulfonate, TRC
Artikelnummer. 30011522
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011522 1 g
30005860 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011522 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2946901G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62319-13-5
Molekylær formel C11H18O4S
Molekylvægt (g/mol) 246.32
Synonym 7,7-Dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic Acid Methyl Ester, (1S,4R)-10-Camphorsulfonic Acid Methyl Ester, (1S)-7,7-dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic Acid Methyl Ester;
IUPAC navn methyl [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonate
SMIL CC1([C@@H]2CC[C@]1(C(=O)C2)CS(=O)(=O)OC)C
Tekniske data
Kemisk navn eller materiale Methyl (1S)-(+)-10-Camphorsulfonate
Mængde 1 g
InChI formel InChI=1S/C11H18O4S/c1-10(2)8-4-5-11(10,9(12)6-8)7-16(13,14)15-3/h8H,4-7H2,1-3H3/t8-,11-/m1/s1
Formel vægt 246.0926