Learn More
Methyl (1R,3S)-2,2-dimethyl-3-(2-oxopropyl)-cyclopropaneacetate, 96%, Thermo Scientific™
Catalog Number 10185010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
CAS 54878-01-2
Color Brown
Boiling Point 104°C to 106°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C11H18O3
Refractive Index 1.4514 to 1.4534
MDL Number MFCD00144756
Quantity 1 g
Specific Rotation -26.6
Synonym methyl 2-1r,3s-2,2-dimethyl-3-2-oxopropyl cyclopropyl acetate, methyl 1r,3s-2,2-dimethyl-3-2-oxo-propyl cyclopropyl acetate, methyl 1 r,3 s 2-2 dimethyl-3-oxopropyl-cyclopropaneacetate, ?-2,2-dimethyl-3beta-2-oxopropyl cyclopropane-1beta-acetic acid methyl ester
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,benzene,petroleum ether,soluble in ethanol,methanol,ccl4
InChI Key MGHKLYIYPFGGDY-DTWKUNHWSA-N
SMILES CC(=O)CC1C(C1(C)C)CC(=O)OC
IUPAC Name methyl 2-[(1R,3S)-2,2-dimethyl-3-(2-oxopropyl)cyclopropyl]acetate
Molecular Weight (g/mol) 198.26
PubChem CID 736111
Formula Weight 198.26
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl (1R, 3S)-2, 2-dimethyl-3-(2-oxopropyl)-cyclopropaneacetate, 96%