Få mere at vide
Methyclothiazide, LoGiCal
Artikelnummer. 30458261
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458261 10 mg
1 options
Artikelnummer. 30458261 Leverandør LoGiCal Leverandørnr. LGCFOR1334.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyclothiazide
CAS 135-07-9
Analyt- eller komponentnavne Methyclothiazide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H11 Cl2 N3 O4 S2
Certificeringer/compliance ISO 9001
Synonym Methyclothiazide, Methyclothiazid, 6-chloro-3-(chloromethyl)-3,4-dihydro-2-methyl-2H-1,2,4-benzothiadiazine-7-sulfonamide-1,1-dioxide, 6-Chloro-3-chloromethyl-3,4-dihydro-2-methyl-7-sulfamoyl-1,2,4-benzothiadiazine 1,1-dioxide, Aquatensen, Ciba 7272-Su, Duretic, Enduron, Enduronum, Eutron, Methyclothiazid, Methyclothiazide, Methycyclothiazide, Methylcyclothiazide, NSC 110431, Naturon
InChI formel InChI=1S/C9H11Cl2N3O4S2/c1-14-9(4-10)13-6-2-5(11)7(19(12,15)16)3-8(6)20(14,17)18/h2-3,9,13H,4H2,1H3,(H2,12,15,16)
Løsningstype Neat
SMIL CN1C(CCl)Nc2cc(Cl)c(cc2S1(=O)=O)S(=O)(=O)N
IUPAC navn 6-chloro-3-(chloromethyl)-2-methyl-1,1-dioxo-3,4-dihydro-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 360.24
Mængde 10 mg
Formel vægt 358.9568 g/mol