Få mere at vide
Methsuximide-d5, TRC
Artikelnummer. 30466844
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30466844 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466844 Leverandør Toronto Research Chemicals Leverandørnr. TRCM27190210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methsuximide-d5
CAS 1189980-63-9
Renhedsgrad noter HPLC
Molekylær formel C12H8D5NO2
Mængde 10 mg
InChI formel InChI=1S/C12H13NO2/c1-12(9-6-4-3-5-7-9)8-10(14)13(2)11(12)15/h3-7H,8H2,1-2H3/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2(C)CC(=O)N(C)C2=O
Molekylvægt (g/mol) 208.27
Procent renhed >95