Få mere at vide
Methoxypromazine Maleate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methoxypromazine Maleate |
| CAS | 3403-42-7 |
| Analyt- eller komponentnavne | Methoxypromazine Maleate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H22 N2 O S . C4 H4 O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine-10-propanamine, 2-methoxy-N,N-dimethyl-, (2Z)-2-butenedioate (1:1), 10H-Phenothiazine-10-propanamine, 2-methoxy-N,N-dimethyl-, (Z)-2-butenedioate (1:1), Phenothiazine, 10-[3-(dimethylamino)propyl]-2-methoxy-, maleate (1:1) (8CI), 2-Methoxypromazine maleate, Methopromazine maleate, Methoxypromazine maleate, Metopromazine maleate, Mopazine, Mopazine maleate, NSC 169469, Tentone, Tentone maleate, Vetomazine |
| InChI formel | InChI=1S/C18H22N2OS.C4H4O4/c1-19(2)11-6-12-20-15-7-4-5-8-17(15)22-18-10-9-14(21-3)13-16(18)20;5-3(6)1-2-4(7)8/h4-5,7-10,13H,6,11-12H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Løsningstype | Neat |
| SMIL | COc1ccc2Sc3ccccc3N(CCCN(C)C)c2c1.OC(=O)\C=C/C(=O)O |
| Vis mere |