Få mere at vide
Methoxyl-d3-amine Hydrochloride, TRC
Artikelnummer. 30476175
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476175 250 mg
30469557 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476175 Leverandør Toronto Research Chemicals Leverandørnr. TRCM262722250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110220-55-8
Molekylær formel C 2H3 H2 N O . Cl H
Molekylvægt (g/mol) 86.54
Synonym Hydroxylamine, O-(methyl-d3)-, hydrochloride (9CI), (Aminooxy)methane-d3 hydrochloride, Hydroxylamine methyl-d3 ether hydrochloride, Methoxy-d3-amine hydrochloride, Methoxylamine-d3 hydrochloride, Methoxyl-d3-amine hydrochloride, N-Methoxylamine-d3 hydrochloride, O-(Methyl-d3)hydroxylamine hydrochloride, O-Methylhydroxylamine-D3 hydrochloride
IUPAC navn O-(trideuteriomethyl)hydroxylamine;hydrochloride
SMIL Cl.[2H]C([2H])([2H])ON
Tekniske data
Kemisk navn eller materiale Methoxyl-d3-amine Hydrochloride
Mængde 250 mg
InChI formel InChI=1S/CH5NO.ClH/c1-3-2;/h2H2,1H3;1H/i1D3;
Formel vægt 86.0326