Få mere at vide
Methoxyl-d3-amine HCl, CDN
Artikelnummer. 30321900
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321900 0.25 g -
30322180 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321900 Leverandør CDN Leverandørnr. CDND12720.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methoxyl-d3-amine Hydrochloride
CAS 110220-55-8
Molekylær formel C 2H3 H2 N O . Cl H
Mængde 0.25 g
InChI formel InChI=1 S/CH5NO.ClH/c1-3-2;/h2H2,1H3;1 H/i1D3;
SMIL Cl.[2 H]C([2 H])([2 H])ON
IUPAC navn O-(trideuteriomethyl)hydroxylamine;hydrochloride
Molekylvægt (g/mol) 86.54
Formel vægt 86.0326 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity