missing translation for 'onlineSavingsMsg'
Få mere at vide

α-(Methoxyimino)-2-furanacetyl Chloride, TRC

Product Code. 30469989
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30469989 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469989 Supplier Toronto Research Chemicals Supplier No. TRCM2685701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale alpha-Z-(Methoxyimino)-2-furanacetyl Chloride
CAS 59176-08-8
Molekylær formel C7H6ClNO3
Mængde 1 g
Synonym (αZ)-α-(Methoxyimino)-2-furanacetyl chloride, (2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl chloride
InChI formel InChI=1S/C7H6ClNO3/c1-11-9-6(7(8)10)5-3-2-4-12-5/h2-4H,1H3/b9-6-
SMIL ClC(/C(C1=CC=CO1)=N\OC)=O
IUPAC navn (Z)-2-(furan-2-yl)-2-(methoxyimino)acetyl chloride
Molekylvægt (g/mol) 187.58
Formel vægt 187
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.