Få mere at vide
Methoxygonadiene (>90%), TRC
Artikelnummer. 30133020
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133020 10 g
30010254 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133020 Leverandør Toronto Research Chemicals Leverandørnr. TRCM19860010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2322-77-2
Molekylær formel C20 H28 O2
Molekylvægt (g/mol) 300.44
Synonym Gona-2,5(10)-dien-17-one, 13-ethyl-3-methoxy-, Levonorgestrel Imp. R, 13-Ethyl-3-methoxygona-2,5(10)-dien-17-one
IUPAC navn (8R,9S,13S,14S)-13-ethyl-3-methoxy-4,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-one
SMIL CC[C@]12CC[C@H]3[C@@H](CCC4=C3CC=C(C4)OC)[C@@H]1CCC2=O
Tekniske data
Kemisk navn eller materiale Methoxygonadiene
Mængde 10 g
InChI formel InChI=1S/C20H28O2/c1-3-20-11-10-16-15-7-5-14(22-2)12-13(15)4-6-17(16)18(20)8-9-19(20)21/h5,16-18H,3-4,6-12H2,1-2H3/t16-,17-,18+,20+/m1/s1
Formel vægt 300.2089